Assay Details
| Gene Symbol: Vps16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030559.3 | NP_085036.3 | 4 - 5 | 440 | 58 | |
| GenBank mRNA | AB056721.1 | - | 4 - 5 | 398 | 58 | |
| AK049872.1 | - | 4 - 5 | 397 | 58 | ||
| AK162984.1 | - | 4 - 5 | 407 | 58 | ||
| BC025626.1 | - | 4 - 5 | 406 | 5320572 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vps16
Genomic Map