Assay Details
| Gene Symbol: Gabpb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271467.1 | NP_001258396.1 | 6 - 7 | 920 | 135 | |
| NM_001271468.1 | NP_001258397.1 | 5 - 6 | 822 | 135 | ||
| NM_001271470.1 | NP_001258399.1 | 6 - 7 | 987 | 135 | ||
| NM_001271492.1 | NP_001258421.1 | 5 - 6 | 822 | 135 | ||
| NM_010249.2 | NP_034379.1 | 5 - 6 | 822 | 135 | ||
| NM_207669.2 | NP_997552.1 | 5 - 6 | 822 | 135 | ||
| GenBank mRNA | AK040722.1 | - | 5 - 6 | 823 | 135 | |
| AK052828.1 | - | 798 | 135 | |||
| AK139710.1 | - | 6 - 7 | 987 | 135 | ||
| AK140222.1 | - | 6 - 7 | 873 | 135 | ||
| AK140907.1 | - | 5 - 6 | 733 | 135 | ||
| AY282800.1 | - | 2 - 3 | 279 | 135 | ||
| AY282802.1 | - | 3 - 4 | 474 | 135 | ||
| AY282803.1 | - | 2 - 3 | 279 | 135 | ||
| BC013558.1 | - | 5 - 6 | 713 | 3592119 | 135 | |
| CA552206.1 | - | 2 - 3 | 239 | 135 | ||
| M74516.1 | - | 5 - 6 | 717 | 135 | ||
| M74517.1 | - | 5 - 6 | 716 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gabpb1
Genomic Map