Assay Details
Gene Symbol: Usp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252580.1 | NP_001239509.1 | 11 - 12 | 2038 | 69 | |
NM_019729.3 | NP_062703.2 | 11 - 12 | 2005 | 69 | ||
GenBank mRNA | AB045709.1 | - | 10 - 11 | 1779 | 69 | |
AF057146.1 | - | 10 - 11 | 1796 | 69 | ||
AK122195.1 | - | 11 - 12 | 1916 | 69 | ||
AK129003.1 | - | 11 - 12 | 2006 | 69 | ||
AK134058.1 | - | 11 - 12 | 1960 | 69 | ||
AK142881.1 | - | 8 - 9 | 1503 | 69 | ||
AK153015.1 | - | 10 - 11 | 1753 | 69 | ||
AK166540.1 | - | 9 - 10 | 1359 | 69 | ||
BC027052.1 | - | 1 - 2 | 558 | 69 | ||
BC050947.1 | - | 11 - 12 | 1970 | 6825981 | 69 | |
BC061465.1 | - | 11 - 12 | 1970 | 6825981 | 69 | |
BC066126.1 | - | 11 - 12 | 1968 | 30301665 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
