Assay Details
| Gene Symbol: Il13ra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089687.2 | - | 8 - 9 | 1364 | 130 | |
| BC003723.1 | - | 5 - 6 | 540 | 3486223 | 130 | |
| EF219410.1 | - | 3 - 4 | 379 | 130 | ||
| U65747.1 | - | 5 - 6 | 628 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il13ra2
Genomic Map