Assay Details
| Gene Symbol: Il13ra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089687.2 | - | 11 - 12 | 1818 | 103 | |
| BC003723.1 | - | 8 - 9 | 994 | 3486223 | 103 | |
| EF219410.1 | - | 6 - 7 | 833 | 103 | ||
| U65747.1 | - | 8 - 9 | 1082 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il13ra2
Genomic Map