Assay Details
| Gene Symbol: Gcnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008105.3 | NP_032131.2 | 1 - 2 | 1529 | 60 | |
| GenBank mRNA | AB037597.1 | - | 1 - 2 | 920 | 60 | |
| AK054226.1 | - | 1 - 2 | 1044 | 60 | ||
| AK080061.1 | - | 1 - 2 | 1095 | 60 | ||
| AY236873.1 | - | 1 - 2 | 1404 | 60 | ||
| AY435149.1 | - | 1 - 2 | 920 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcnt2
Genomic Map