Assay Details
| Gene Symbol: Nedd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111324.2 | NP_001104794.1 | 6 - 7 | 2251 | 70 | |
| NM_017464.5 | NP_059492.3 | 5 - 6 | 2032 | 70 | ||
| GenBank mRNA | AF009366.1 | - | 6 - 7 | 2187 | 70 | |
| AK030985.1 | - | 6 - 7 | 2185 | 70 | ||
| AK033729.1 | - | 5 - 6 | 2033 | 70 | ||
| AK046357.1 | - | 5 - 6 | 1995 | 70 | ||
| AK054179.1 | - | 5 - 6 | 2028 | 70 | ||
| AK083374.1 | - | 6 - 7 | 2179 | 70 | ||
| BC004696.1 | - | 1 - 2 | 300 | 70 | ||
| BC053713.1 | - | 6 - 7 | 2184 | 30077589 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nedd9
Genomic Map