Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300003154.mm.1 |
NM_001205043.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003154.mm.1 |
NM_001205043.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003154.mm.1 |
NM_001205044.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003154.mm.1 |
NM_001205044.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003154.mm.1 |
NM_021878.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003154.mm.1 |
NM_021878.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003154.mm.1 |
TC1300000514.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300003155.mm.1 |
NM_001205043.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003155.mm.1 |
NM_001205043.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003155.mm.1 |
NM_001205044.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003155.mm.1 |
NM_001205044.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003155.mm.1 |
NM_021878.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003155.mm.1 |
NM_021878.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300003155.mm.1 |
TC1300000514.mm.1 |
MTA TC |