Assay Details
| Gene Symbol: Siglech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178706.4 | NP_848821.2 | 4 - 5 | 972 | 65 | |
| GenBank mRNA | AK032377.1 | - | 3 - 4 | 877 | 65 | |
| AK045719.1 | - | 5 - 6 | 602 | 65 | ||
| AK047813.1 | - | 4 - 5 | 897 | 65 | ||
| AK080658.1 | - | 3 - 4 | 687 | 65 | ||
| BC098229.1 | - | 4 - 5 | 911 | 1430130 | 65 | |
| BC120501.1 | - | 4 - 5 | 914 | 40129424 | 65 | |
| BC144741.1 | - | 3 - 4 | 635 | 9053289 | 65 | |
| BC144743.1 | - | 4 - 5 | 914 | 9053291 | 65 | |
| DQ157764.1 | - | 4 - 5 | 891 | 65 | ||
| DQ208935.1 | - | 4 - 5 | 917 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Siglech
Genomic Map