Assay Details
Gene Symbol: Aloxe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011786.2 | NP_035916.2 | 4 - 5 | 575 | 76 | |
GenBank mRNA | AK141003.1 | - | 4 - 5 | 738 | 76 | |
BC116258.1 | - | 4 - 5 | 574 | 40093733 | 76 | |
BC116259.1 | - | 4 - 5 | 574 | 40093735 | 76 | |
Y14695.1 | - | 4 - 5 | 575 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
