Assay Details
| Gene Symbol: Slc39a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012305.2 | NP_001012305.1 | 8 - 9 | 1595 | 141 | |
| GenBank mRNA | AK134622.1 | - | 8 - 9 | 1596 | 141 | |
| BC089362.1 | - | 8 - 9 | 1589 | 30628112 | 141 | |
| BC113764.1 | - | 8 - 9 | 1578 | 40059402 | 141 | |
| BC113765.1 | - | 8 - 9 | 1578 | 40059404 | 141 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc39a12
Genomic Map