Assay Details
| Gene Symbol: Alox12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009659.2 | NP_033789.1 | 12 - 13 | 1833 | 99 | |
| GenBank mRNA | AF059251.1 | - | 12 - 13 | 1711 | 99 | |
| BC113149.1 | - | 12 - 13 | 1797 | 40054352 | 99 | |
| Y14334.1 | - | 12 - 13 | 1833 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alox12b
Genomic Map