Assay Details
Gene Symbol: Rreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039188.1 | NP_001034277.1 | 3 - 4 | 548 | 78 | |
NM_001177868.1 | NP_001171339.1 | 3 - 4 | 273 | 78 | ||
NM_001177869.1 | NP_001171340.1 | 2 - 3 | 209 | 78 | ||
NM_026830.2 | NP_081106.1 | 3 - 4 | 516 | 78 | ||
NR_033218.1 | 3 - 4 | 456 | 78 | |||
NR_033615.1 | 3 - 4 | 273 | 78 | |||
GenBank mRNA | AK047047.1 | - | 3 - 4 | 253 | 78 | |
AK047251.1 | - | 3 - 4 | 253 | 78 | ||
AK147260.1 | - | 3 - 4 | 457 | 78 | ||
AK147653.1 | - | 3 - 4 | 457 | 78 | ||
AK154980.1 | - | 3 - 4 | 516 | 78 | ||
AK170652.1 | - | 3 - 4 | 548 | 78 | ||
AK171375.1 | - | 3 - 4 | 253 | 78 | ||
BC080680.1 | - | 3 - 4 | 362 | 6330949 | 78 | |
BY206933.1 | - | 3 - 4 | 311 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK147260.1, AK147653.1, AK154980.1, AK171375.1, BC080680.1, NM_001039188.1, NM_001177868.1, NM_001177869.1, NM_026830.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
