Assay Details
| Gene Symbol: Usp6nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080548.1 | NP_001074017.1 | 7 - 8 | 661 | 80 | |
| NM_181399.3 | NP_852064.2 | 6 - 7 | 500 | 80 | ||
| GenBank mRNA | AK084045.1 | - | 7 - 8 | 484 | 80 | |
| AK129037.1 | - | 7 - 8 | 513 | 80 | ||
| AK155073.1 | - | 7 - 8 | 662 | 80 | ||
| BC051184.1 | - | 6 - 7 | 503 | 80 | ||
| BQ938063.1 | - | 6 - 7 | 500 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp6nl
Genomic Map