Assay Details
Gene Symbol: Acsl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033600.1 | NP_001028772.1 | 5 - 6 | 809 | 111 | |
NM_019477.3 | NP_062350.3 | 5 - 6 | 812 | 111 | ||
NM_207625.2 | NP_997508.1 | 5 - 6 | 1102 | 111 | ||
GenBank mRNA | AB033885.1 | - | 5 - 6 | 621 | 111 | |
AB033886.1 | - | 5 - 6 | 911 | 111 | ||
AB033887.1 | - | 5 - 6 | 959 | 111 | ||
AJ243502.1 | - | 5 - 6 | 657 | 111 | ||
AK054387.1 | - | 5 - 6 | 782 | 111 | ||
BC016416.1 | - | 5 - 6 | 683 | 3157957 | 111 | |
BC058663.1 | - | 5 - 6 | 898 | 6409748 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
