Assay Details
| Gene Symbol: Gucy2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007576.2 | NP_001007577.1 | 13 - 14 | 2824 | 111 | |
| GenBank mRNA | AY651761.1 | - | 13 - 14 | 2819 | 111 | |
| BC115714.1 | - | 12 - 13 | 2654 | 40085764 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gucy2f
Genomic Map