Assay Details
| Gene Symbol: Gucy2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007576.2 | NP_001007577.1 | 9 - 10 | 2206 | 131 | |
| GenBank mRNA | AK044234.1 | - | 9 - 10 | 2207 | 131 | |
| AY651761.1 | - | 9 - 10 | 2201 | 131 | ||
| BC115714.1 | - | 8 - 9 | 2036 | 40085764 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gucy2f
Genomic Map