Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200008202.mm.1 |
NR_028261.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008202.mm.1 |
NR_028261.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008202.mm.1 |
TC1200001142.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008195.mm.1 |
NR_028261.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008195.mm.1 |
NR_028261.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008195.mm.1 |
TC1200001142.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008194.mm.1 |
NR_028261.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008194.mm.1 |
NR_028261.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008194.mm.1 |
TC1200001142.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008209.mm.1 |
NR_028261.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008209.mm.1 |
NR_028261.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008209.mm.1 |
TC1200001142.mm.1 |
MTA TC |