Assay Details
| Gene Symbol: Sprr2j-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003185.1 | 1 - 2 | 80 | 83 | ||
| GenBank mRNA | AK019074.1 | - | 1 - 2 | 82 | 83 | |
| AY158994.1 | - | 1 - 2 | 85 | 83 | ||
| BC125542.1 | - | 1 - 2 | 56 | 83 | ||
| BC125544.1 | - | 1 - 2 | 56 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small proline-rich protein 2I | Sprr2i | -8.8kb | Sense | Chr. 3 - 92407991 - 92409271 on NCBI Build GRCm38 | |
| Downstream | small proline-rich protein 2K | Sprr2k | 13.2kb | Sense | Chr. 3 - 92432582 - 92433927 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK019074.1, AY158994.1 transcripts.
Set Membership: |
|

Sprr2j-ps
Genomic Map