Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600004563.mm.1 |
TC0600000591.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004563.mm.1 |
NM_001164733.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004563.mm.1 |
NM_001164733.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004563.mm.1 |
NM_001164734.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004563.mm.1 |
NM_001164734.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004563.mm.1 |
NM_019939.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004563.mm.1 |
NM_019939.2 |
Refseq |