Assay Details
Gene Symbol: Mpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164733.1 | NP_001158205.1 | 7 - 8 | 969 | 115 | |
NM_001164734.1 | NP_001158206.1 | 7 - 8 | 979 | 115 | ||
GenBank mRNA | AF199010.1 | - | 7 - 8 | 975 | 115 | |
AK144434.1 | - | 7 - 8 | 980 | 115 | ||
AK164549.1 | - | 7 - 8 | 970 | 115 | ||
BC145363.1 | - | 7 - 8 | 837 | 9053921 | 115 | |
BC145364.1 | - | 7 - 8 | 827 | 9053922 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
