Assay Details
| Gene Symbol: Duoxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025777.2 | NP_080053.1 | 2 - 3 | 345 | 73 | |
| GenBank mRNA | AK018569.1 | - | 2 - 3 | 347 | 73 | |
| BC031111.1 | - | 2 - 3 | 332 | 4989815 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Duoxa2
Genomic Map