Assay Details
Gene Symbol: Patl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026251.2 | NP_080527.2 | 10 - 11 | 1260 | 60 | |
GenBank mRNA | AK010811.1 | - | 3 - 4 | 332 | 60 | |
BC145646.1 | - | 8 - 9 | 1041 | 40130949 | 60 | |
DV660077.1 | - | 1 - 2 | 124 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
