Assay Details
Gene Symbol: Patl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026251.2 | NP_080527.2 | 14 - 15 | 1705 | 67 | |
GenBank mRNA | AK010811.1 | - | 7 - 8 | 67 | ||
AK015195.1 | - | 14 - 15 | 1709 | 67 | ||
BC145646.1 | - | 12 - 13 | 1486 | 40130949 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
