Assay Details
Gene Symbol: Patl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026251.2 | NP_080527.2 | 15 - 16 | 1854 | 95 | |
GenBank mRNA | AK010811.1 | - | 8 - 9 | 922 | 95 | |
AK015195.1 | - | 15 - 16 | 1858 | 95 | ||
BC145646.1 | - | 13 - 14 | 1635 | 40130949 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
