Assay Details
Gene Symbol: Etl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029895.4 | NP_084171.2 | 17 - 18 | 3962 | 95 | |
NM_178059.5 | NP_835160.2 | 17 - 18 | 3995 | 95 | ||
GenBank mRNA | AB125594.2 | - | 17 - 18 | 3995 | 95 | |
AB125595.2 | - | 17 - 18 | 3962 | 95 | ||
BC026657.1 | - | 5 - 6 | 924 | 95 | ||
BC172143.1 | - | 16 - 17 | 3500 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
