Assay Details
Gene Symbol: Etl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081006.1 | NP_001074475.1 | 6 - 7 | 1090 | 70 | |
NM_001177630.2 | NP_001171101.2 | 5 - 6 | 1188 | 70 | ||
NM_001177631.1 | NP_001171102.1 | 5 - 6 | 1188 | 70 | ||
NM_029895.4 | NP_084171.2 | 6 - 7 | 1401 | 70 | ||
NM_178059.5 | NP_835160.2 | 6 - 7 | 1401 | 70 | ||
GenBank mRNA | AB093289.2 | - | 5 - 6 | 978 | 70 | |
AB125594.2 | - | 6 - 7 | 1401 | 70 | ||
AB125595.2 | - | 6 - 7 | 1401 | 70 | ||
AK052020.1 | - | 7 - 8 | 1503 | 70 | ||
AK147510.1 | - | 5 - 6 | 1095 | 70 | ||
AK158471.1 | - | 1 - 2 | 136 | 70 | ||
BC158051.1 | - | 5 - 6 | 910 | 9007405 | 70 | |
BC172143.1 | - | 5 - 6 | 908 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
