Assay Details
| Gene Symbol: Etl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081006.1 | NP_001074475.1 | 8 - 9 | 2025 | 86 | |
| NM_001177630.2 | NP_001171101.2 | 7 - 8 | 2123 | 86 | ||
| GenBank mRNA | BC158051.1 | - | 7 - 8 | 1845 | 9007405 | 86 |
More Information
Set Membership: |
|

Etl4
Genomic Map