Assay Details
| Gene Symbol: Ston2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175367.6 | NP_780576.1 | 4 - 5 | 1057 | 116 | |
| GenBank mRNA | AK036612.1 | - | 4 - 5 | 1058 | 116 | |
| BC132531.1 | - | 3 - 4 | 643 | 40130808 | 116 | |
| BC138054.1 | - | 3 - 4 | 643 | 8861073 | 116 | |
More Information
Set Membership: |
|

Ston2
Genomic Map