Assay Details
| Gene Symbol: Aqp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007472.2 | NP_031498.1 | 2 - 3 | 737 | 57 | |
| GenBank mRNA | AK081886.1 | - | 2 - 3 | 605 | 57 | |
| AK086688.1 | - | 2 - 3 | 605 | 57 | ||
| AK128905.1 | - | 2 - 3 | 604 | 57 | ||
| AK157333.1 | - | 2 - 3 | 605 | 57 | ||
| AK158226.1 | - | 2 - 3 | 605 | 57 | ||
| AK158389.1 | - | 2 - 3 | 605 | 57 | ||
| AK166280.1 | - | 2 - 3 | 605 | 57 | ||
| AK171627.1 | - | 2 - 3 | 605 | 57 | ||
| AK172361.1 | - | 2 - 3 | 605 | 57 | ||
| L02914.1 | - | 2 - 3 | 604 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aqp1
Genomic Map