Assay Details
| Gene Symbol: Znrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199143.2 | NP_954594.1 | 2 - 3 | 652 | 118 | |
| GenBank mRNA | AF513708.1 | - | 2 - 3 | 652 | 118 | |
| BC104316.2 | - | 2 - 3 | 297 | 118 | ||
| BC104317.2 | - | 2 - 3 | 306 | 118 | ||
| BC125242.1 | - | 2 - 3 | 306 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC104316.2, BC104317.2, BC125242.1 transcripts.
Set Membership: |
|

Znrf2
Genomic Map