Assay Details
| Gene Symbol: Ghrhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003685.3 | NP_001003685.2 | 10 - 11 | 1069 | 67 | |
| GenBank mRNA | AK030504.1 | - | 10 - 11 | 1022 | 67 | |
| BC120748.1 | - | 10 - 11 | 999 | 40129671 | 67 | |
| BC120774.1 | - | 10 - 11 | 999 | 40129697 | 67 | |
| L07379.1 | - | 10 - 11 | 1002 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ghrhr
Genomic Map