Assay Details
| Gene Symbol: Plekha8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001335.2 | NP_001001335.1 | 3 - 4 | 546 | 80 | |
| NM_001164361.1 | NP_001157833.1 | 3 - 4 | 641 | 80 | ||
| GenBank mRNA | AK078040.1 | - | 3 - 4 | 642 | 80 | |
| AK170592.1 | - | 3 - 4 | 408 | 80 | ||
| BC052360.1 | - | 3 - 4 | 546 | 6704789 | 80 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Plekha8
Genomic Map