Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600005020.mm.1 |
TC0600000655.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600005020.mm.1 |
NM_001001335.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005020.mm.1 |
NM_001001335.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005020.mm.1 |
NM_001164361.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005020.mm.1 |
NM_001164361.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005019.mm.1 |
TC0600000655.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600005019.mm.1 |
NM_001001335.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005019.mm.1 |
NM_001001335.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005019.mm.1 |
NM_001164361.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600005019.mm.1 |
NM_001164361.1 |
Refseq |