Assay Details
Gene Symbol: Med22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033908.1 | NP_001029080.1 | 1 - 2 | 346 | 74 | |
NM_011513.2 | NP_035643.1 | 1 - 2 | 346 | 74 | ||
GenBank mRNA | AK144744.1 | - | 1 - 2 | 325 | 74 | |
AK161053.1 | - | 1 - 2 | 287 | 74 | ||
AK166250.1 | - | 1 - 2 | 292 | 74 | ||
AK166474.1 | - | 1 - 2 | 319 | 74 | ||
BC018225.1 | - | 1 - 2 | 336 | 3968506 | 74 | |
BG805496.1 | - | 1 - 2 | 208 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081083 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081082 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081081 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081079 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081080 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P141554 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 118075 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385587 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 911983 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 836349 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 792340 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140008 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P141554 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081080 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081045 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081054 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081060 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081057 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081079 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081059 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081066 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081041 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081083 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081064 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081073 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081042 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081047 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081040 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081063 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081061 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081044 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081048 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081072 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081058 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081074 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081043 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081068 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081046 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081051 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081049 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21081077 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140008 | - | - |
Whole Mouse Genome G4122A | A_51_P141554 | - | - |
Whole Mouse Genome G4122A | A_52_P17194 | BC018225.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P17194 | BG805496.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P17194 | AK166250.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P17194 | AK166474.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P17194 | AK144744.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133916 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140008 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01411196 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01192531 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081081 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081079 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081082 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081083 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21081080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21080995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21080994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21080999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21080998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21081066 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081047 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081068 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01192535 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081083 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01575190 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01575171 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081045 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081077 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081046 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01913980 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081040 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081075 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081038 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081079 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081051 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081042 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081066 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081060 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081064 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081061 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01575184 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01575178 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21081063 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01575194 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10481133 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481132 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481130 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481129 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481134 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481131 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1421528_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10481128 | - | - |
Mouse Genome 430 2.0 Array | 1421528_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1421528_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20933_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20933_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200026912.mm.1 | NM_001033908.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200026912.mm.1 | NM_001033908.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200026912.mm.1 | NM_011513.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200026912.mm.1 | NM_011513.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200026912.mm.1 | TC0200003203.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200026910.mm.1 | NM_001033908.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200026910.mm.1 | NM_001033908.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200026910.mm.1 | NM_011513.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200026910.mm.1 | NM_011513.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200026910.mm.1 | TC0200003203.mm.1 | MTA TC |
Mouse Expression Set 430 | 1421528_PM_a_at | - | - |
Mouse Expression Set 430 | 1421528_a_at | - | - |
Murine Genome U74v2 Set | 98430_at | - | - |
Murine Genome U74v2 Set | 98430_PM_at | - | - |
Murine Genome U74v2 Set | 161561_PM_r_at | - | - |
Murine Genome U74v2 Set | 161561_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1421528_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1421528_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421528_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421528_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3290722 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003290722 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3290722 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl19515.3_120-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3290722 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3041056 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3100456 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3041056 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3100456 | - | - |
