Assay Details
| Gene Symbol: Abo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290444.1 | NP_001277373.1 | 1 - 2 | 121 | 76 | |
| NM_030718.5 | NP_109643.3 | 1 - 2 | 121 | 76 | ||
| GenBank mRNA | AK033786.1 | - | 1 - 2 | 42 | 76 | |
| AK035261.1 | - | 1 - 2 | 106 | 76 | ||
| BC116759.1 | - | 1 - 2 | 70 | 40126078 | 76 | |
| BC116761.1 | - | 1 - 2 | 70 | 40126080 | 76 | |
| DV046627.1 | - | 1 - 2 | 140 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033786.1, AK035261.1, BC116759.1, BC116761.1, NM_001290444.1, NM_030718.5 transcripts.
Set Membership: |
|

Abo
Genomic Map