Assay Details
| Gene Symbol: Ddx46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282055.1 | NP_001268984.1 | 10 - 11 | 1518 | 79 | |
| GenBank mRNA | AK088950.1 | - | 10 - 11 | 1490 | 79 | |
| AK129220.1 | - | 10 - 11 | 1491 | 79 | ||
| BC026492.1 | - | 3 - 4 | 463 | 4505095 | 79 | |
| BC092240.1 | - | 9 - 10 | 1429 | 30662573 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx46
Genomic Map