Assay Details
| Gene Symbol: Uimc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011307.2 | NP_035437.1 | 4 - 5 | 461 | 64 | |
| NR_121592.1 | 4 - 5 | 466 | 64 | |||
| GenBank mRNA | AK052704.1 | - | 4 - 5 | 462 | 64 | |
| AK083342.1 | - | 4 - 5 | 402 | 64 | ||
| BC037092.1 | - | 4 - 5 | 422 | 64 | ||
| BC040808.1 | - | 4 - 5 | 417 | 4038778 | 64 | |
| BC051659.1 | - | 4 - 5 | 384 | 64 | ||
| BC066220.1 | - | 4 - 5 | 384 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uimc1
Genomic Map