Assay Details
| Gene Symbol: Pde11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081033.1 | NP_001074502.1 | 12 - 13 | 2093 | 63 | |
| GenBank mRNA | AK042383.1 | - | 9 - 10 | 871 | 63 | |
| BC158125.1 | - | 12 - 13 | 2233 | 9088197 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pde11a
Genomic Map