Assay Details
| Gene Symbol: Pde11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081033.1 | NP_001074502.1 | 18 - 19 | 2610 | 70 | |
| GenBank mRNA | BC158125.1 | - | 18 - 19 | 2750 | 9088197 | 70 |
More Information
Set Membership: |
|

Pde11a
Genomic Map