Assay Details
| Gene Symbol: Slc27a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011989.4 | NP_036119.1 | 5 - 6 | 1070 | 65 | |
| GenBank mRNA | AF072759.1 | - | 3 - 4 | 372 | 65 | |
| AJ251113.1 | - | 4 - 5 | 783 | 65 | ||
| AK036919.1 | - | 5 - 6 | 1072 | 65 | ||
| AK155313.1 | - | 5 - 6 | 1039 | 65 | ||
| AK155388.1 | - | 5 - 6 | 1040 | 65 | ||
| AK170377.1 | - | 5 - 6 | 1074 | 65 | ||
| BC023114.1 | - | 5 - 6 | 921 | 4158956 | 65 | |
| CT010312.1 | - | 4 - 5 | 783 | 65 | ||
More Information
Set Membership: |
|

Slc27a4
Genomic Map