Assay Details
| Gene Symbol: Slc27a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011989.4 | NP_036119.1 | 6 - 7 | 1164 | 81 | |
| GenBank mRNA | AJ251113.1 | - | 5 - 6 | 877 | 81 | |
| AK036919.1 | - | 6 - 7 | 1166 | 81 | ||
| AK155313.1 | - | 6 - 7 | 1133 | 81 | ||
| AK155388.1 | - | 6 - 7 | 1134 | 81 | ||
| AK170377.1 | - | 6 - 7 | 1168 | 81 | ||
| BC023114.1 | - | 6 - 7 | 1015 | 4158956 | 81 | |
| CT010312.1 | - | 5 - 6 | 877 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc27a4
Genomic Map