Assay Details
| Gene Symbol: Slc27a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011989.4 | NP_036119.1 | 8 - 9 | 1479 | 67 | |
| GenBank mRNA | AF072759.1 | - | 6 - 7 | 781 | 67 | |
| AJ251113.1 | - | 7 - 8 | 1192 | 67 | ||
| AK009292.1 | - | 2 - 3 | 266 | 67 | ||
| AK033519.1 | - | 4 - 5 | 423 | 67 | ||
| AK036919.1 | - | 8 - 9 | 1481 | 67 | ||
| AK155313.1 | - | 8 - 9 | 1448 | 67 | ||
| AK155388.1 | - | 8 - 9 | 1449 | 67 | ||
| AK170377.1 | - | 8 - 9 | 1483 | 67 | ||
| BC023114.1 | - | 8 - 9 | 1330 | 4158956 | 67 | |
| CT010312.1 | - | 7 - 8 | 1192 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc27a4
Genomic Map