Assay Details
| Gene Symbol: Endog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007931.1 | NP_031957.1 | 1 - 2 | 538 | 71 | |
| GenBank mRNA | AB012108.1 | - | 1 - 2 | 538 | 71 | |
| AK144459.1 | - | 1 - 2 | 570 | 71 | ||
| BC030177.1 | - | 1 - 2 | 547 | 5029633 | 71 | |
| X99394.1 | - | 1 - 2 | 34 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Endog
Genomic Map