Assay Details
| Gene Symbol: Mapkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290625.1 | NP_001277554.1 | 3 - 4 | 497 | 121 | |
| NM_001290626.1 | NP_001277555.1 | 2 - 3 | 495 | 121 | ||
| NM_177345.4 | NP_796319.1 | 3 - 4 | 823 | 121 | ||
| GenBank mRNA | AK042154.1 | - | 2 - 3 | 485 | 121 | |
| AK052045.1 | - | 3 - 4 | 491 | 121 | ||
| AK132263.1 | - | 3 - 4 | 487 | 121 | ||
| AK148428.1 | - | 3 - 4 | 812 | 121 | ||
| BC043296.1 | - | 3 - 4 | 722 | 121 | ||
| BC090644.1 | - | 2 - 3 | 397 | 30629263 | 121 | |
| BC096618.1 | - | 3 - 4 | 796 | 6392317 | 121 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290626.1 transcripts.
Set Membership: |
|

Mapkap1
Genomic Map