Assay Details
Gene Symbol: Erich2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025744.2 | NP_080020.2 | 12 - 13 | 1547 | 150 | |
GenBank mRNA | AK006384.1 | - | 12 - 13 | 875 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549973 | AK017113.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549973 | NM_025744.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549973 | AK006384.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549973 | AK017113.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549973 | NM_025744.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549973 | AK006384.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558898 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 90911 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 854217 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 831414 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72381 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281131 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509532 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899567 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 708429 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665787 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528759 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539023 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789450 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 516764 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 43859 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19157 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33241 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 60242 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549973 | AK017113.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549973 | NM_025744.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549973 | AK006384.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549973 | AK017113.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549973 | NM_025744.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549973 | AK006384.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P50205 | AK016654.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P50205 | AK017113.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P50205 | NM_025744.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P50205 | AK006384.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P549973 | AK017113.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P549973 | NM_025744.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P549973 | AK006384.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P549973 | AK017113.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P549973 | NM_025744.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P549973 | AK006384.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P109119 | AK017113.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116929 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120502 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10472697 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472699 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472706 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472695 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472705 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472696 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472701 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472703 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472704 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472698 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472702 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472700 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1453176_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431907_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10472694 | - | - |
Mouse Genome 430 2.0 Array | 1431907_at | - | - |
Mouse Genome 430 2.0 Array | 1453176_a_at | - | - |
Mouse Genome 430 2.0 Array | 1453176_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1431907_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66748_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66748_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200008173.mm.1 | NM_025744.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200008173.mm.1 | NM_025744.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200008173.mm.1 | TC0200001022.mm.1 | MTA TC |
Mouse Expression Set 430 | 1453176_a_at | - | - |
Mouse Expression Set 430 | 1431907_PM_at | - | - |
Mouse Expression Set 430 | 1431907_at | - | - |
Mouse Expression Set 430 | 1453176_PM_a_at | - | - |
Murine Genome U74v2 Set | 134885_at | - | - |
Murine Genome U74v2 Set | 134885_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431907_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431907_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453176_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453176_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 299865 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 102940692 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2970441 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 102940692 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002970441 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249756 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1224902 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1249756 | - | - |
mouse-6 v1.1 (Illumina) | scl0066748.2_33-S | - | - |
mouse-6 v1.1 (Illumina) | scl20814.13.1_92-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102940692 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2970441 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066748.2_33-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102940692 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2970441 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1249756 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1249756 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0066748.2_33-S | - | - |
