Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200008156.mm.1 |
NM_025744.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008156.mm.1 |
NM_025744.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008156.mm.1 |
TC0200001022.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008157.mm.1 |
NM_025744.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008157.mm.1 |
NM_025744.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008157.mm.1 |
TC0200001022.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008155.mm.1 |
NM_025744.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008155.mm.1 |
NM_025744.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008155.mm.1 |
TC0200001022.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008159.mm.1 |
NM_025744.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008159.mm.1 |
NM_025744.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008159.mm.1 |
TC0200001022.mm.1 |
MTA TC |