Assay Details
Gene Symbol: Kif18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139303.1 | NP_647464.1 | 4 - 5 | 792 | 97 | |
GenBank mRNA | AK036722.1 | - | 4 - 5 | 679 | 97 | |
AK041469.1 | - | 4 - 5 | 691 | 97 | ||
AK044795.1 | - | 4 - 5 | 687 | 97 | ||
AK049197.1 | - | 4 - 5 | 742 | 97 | ||
AK082913.1 | - | 4 - 5 | 705 | 97 | ||
AK135735.1 | - | 4 - 5 | 713 | 97 | ||
AK164649.1 | - | 4 - 5 | 687 | 97 | ||
BC004740.1 | - | 3 - 4 | 647 | 97 | ||
BC016095.1 | - | 4 - 5 | 792 | 4506052 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
