Assay Details
| Gene Symbol: Ccbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK031511.1 | - | 1 - 2 | 185 | 58 | |
| AK042391.1 | - | 1 - 2 | 216 | 58 | ||
| AK088404.1 | - | 1 - 2 | 178 | 58 | ||
| AK163165.1 | - | 1 - 2 | 79 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK042391.1, AK088404.1 transcripts.
Set Membership: |
|

Ccbl1
Genomic Map