Assay Details
| Gene Symbol: Ccbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172404.2 | NP_765992.2 | 3 - 4 | 332 | 66 | |
| GenBank mRNA | AK008165.1 | - | 1 - 2 | 109 | 66 | |
| AK042391.1 | - | 2 - 3 | 278 | 66 | ||
| AK088404.1 | - | 2 - 3 | 240 | 66 | ||
| AK163165.1 | - | 2 - 3 | 141 | 66 | ||
| BC016206.1 | - | 3 - 4 | 434 | 4912335 | 66 | |
| BC025124.1 | - | 2 - 3 | 465 | 4486058 | 66 | |
| BC052047.1 | - | 3 - 4 | 332 | 5687475 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccbl1
Genomic Map